KEY465192688 Key Organics / BIONET

4-(4-methylphenyl)-2-sulfanylidene-1H,2H,3H,4H,5H-indeno[1,2-d]pyrimidin-5-one

  • MDL number MFCD00219297
  • Cas Number : 60477-76-1
  • Molecular Weight : 306.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0309_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart