KEY465192692 Key Organics / BIONET

2-{[1-(4-bromophenyl)-1H-pyrrol-3-yl]methylidene}propanedioic acid

  • MDL number MFCD03102799
  • Cas Number : 860785-98-4
  • Molecular Weight : 336.141

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9664

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0610_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart