KEY465192700 Key Organics / BIONET

N-(2-amino-5-chloro-4-methylphenyl)acetamide

  • MDL number MFCD03011787
  • Cas Number : 383147-42-0
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9678

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0624_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart