KEY465192701 Key Organics / BIONET

N-[5-(3-chlorophenoxy)-4-methyl-2-nitrophenyl]acetamide

  • MDL number MFCD03011788
  • Cas Number : 383147-44-2
  • Molecular Weight : 320.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9679

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0625_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart