KEY465192702 Key Organics / BIONET

1-[1-(2H-1,3-benzodioxol-5-yl)-1H-pyrrol-2-yl]-2,2,2-trichloroethan-1-one

  • MDL number MFCD03011789
  • Cas Number : 383147-46-4
  • Molecular Weight : 332.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9680

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0626_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart