KEY465192706 Key Organics / BIONET

4-[2-(pyridin-2-yl)-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD28347899
  • Cas Number : 1823183-38-5
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 239
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart