KEY465192708 Key Organics / BIONET

5-methyl-2-(methylsulfanyl)-6-[(2E)-3-phenylprop-2-en-1-yl]-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD03305494
  • Cas Number : 860786-00-1
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9704

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0840_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart