KEY465192709 Key Organics / BIONET

(Z)-methoxy({[5-nitro-2-(piperidin-1-yl)phenyl]methylidene})amine

  • MDL number MFCD03012010
  • Cas Number : 383147-50-0
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9711

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-1136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart