KEY465192710 Key Organics / BIONET

(Z)-methoxy({4-[(3-methoxyphenyl)sulfanyl]-3-nitrophenyl}methylidene)amine

  • MDL number MFCD03012011
  • Cas Number : 383147-51-1
  • Molecular Weight : 318.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-1138_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart