KEY465192711 Key Organics / BIONET

2-({4-[(1E)-(methoxyimino)methyl]-2-nitrophenyl}sulfanyl)aniline

  • MDL number MFCD04124519
  • Cas Number : 860786-01-2
  • Molecular Weight : 303.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9713

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-1139_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart