KEY465192715 Key Organics / BIONET

(E)-({2-[(2,6-dichlorophenyl)sulfanyl]-5-nitrophenyl}methylidene)(methoxy)amine

  • MDL number MFCD03012015
  • Cas Number : 383147-56-6
  • Molecular Weight : 357.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-1145_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart