KEY465192722 Key Organics / BIONET

1,3-dimethyl 2-bromo-2-(4-nitrophenyl)propanedioate

  • MDL number MFCD03617781
  • Cas Number : 860786-04-5
  • Molecular Weight : 332.106

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0012_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart