KEY465192723 Key Organics / BIONET

1,3-dimethyl 2-(5-nitropyridin-2-yl)-2-(prop-2-en-1-yl)propanedioate

  • MDL number MFCD03617783
  • Cas Number : 860786-06-7
  • Molecular Weight : 294.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9738

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart