KEY465192725 Key Organics / BIONET

1,3-dimethyl 2-(2-nitrophenyl)-2-(prop-2-en-1-yl)propanedioate

  • MDL number MFCD03617785
  • Cas Number : 610315-36-1
  • Molecular Weight : 293.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9742

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart