KEY465192735 Key Organics / BIONET

ethyl 2-[4-(3-methoxyphenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetate

  • MDL number MFCD03787616
  • Cas Number : 860786-33-0
  • Molecular Weight : 291.307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9780

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0252_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart