KEY465192750 Key Organics / BIONET

methyl 5-bromo-4-(2-chloroacetamido)-2-methoxybenzoate

  • MDL number MFCD03617792
  • Cas Number : 860785-02-0
  • Molecular Weight : 336.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0813_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart