KEY465192759 Key Organics / BIONET

1-{1-[(4-fluorophenyl)methyl]-2-methyl-1H-imidazol-4-yl}ethan-1-one

  • MDL number MFCD03617890
  • Cas Number : 860785-14-4
  • Molecular Weight : 232.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9883

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0838_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart