KEY465192761 Key Organics / BIONET

1-{4-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-2-yl}-4-(naphthalene-2-carbonyl)-1H-1,2,3-triazole

  • MDL number MFCD03787501
  • Cas Number : 860785-20-2
  • Molecular Weight : 355.401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9890

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0854_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart