KEY465192764 Key Organics / BIONET

(2E)-3-{[3-hydroxy-4-(methoxycarbonyl)phenyl]carbamoyl}prop-2-enoic acid

  • MDL number MFCD03787053
  • Cas Number : 871109-58-9
  • Molecular Weight : 265.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9896

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0864_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart