KEY465192770 Key Organics / BIONET

4-[benzyl(ethyl)amino]-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD04124525
  • Cas Number : 860785-50-8
  • Molecular Weight : 310.357

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9934

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart