KEY465192773 Key Organics / BIONET

ethyl 1-ethyl-4-{[(4-methoxyphenyl)methyl]amino}-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD04124528
  • Cas Number : 860785-52-0
  • Molecular Weight : 368.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9941

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart