KEY465192776 Key Organics / BIONET

5-[(1,2-benzoxazol-3-yl)methyl]-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04124534
  • Cas Number : 860785-59-7
  • Molecular Weight : 308.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9949

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0200_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart