KEY465192780 Key Organics / BIONET

2,2,2-trifluoro-1-{1-[(2-fluorophenyl)methyl]-1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one

  • MDL number MFCD04124539
  • Cas Number : 860785-63-3
  • Molecular Weight : 322.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0207_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart