KEY465192786 Key Organics / BIONET

2-(2-carbamimidamido-4-phenyl-1,3-thiazol-5-yl)acetic acid

  • MDL number MFCD04124556
  • Cas Number : 860785-73-5
  • Molecular Weight : 276.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9972

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0243_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart