KEY465192787 Key Organics / BIONET

5-phenylmethanesulfonyl-7-(trifluoromethyl)-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline

  • MDL number MFCD04124559
  • Cas Number : 860786-77-2
  • Molecular Weight : 396.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9975

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0246_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart