KEY465192791 Key Organics / BIONET

8-(trifluoromethyl)-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline

  • MDL number MFCD04124569
  • Cas Number : 860786-84-1
  • Molecular Weight : 242.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Green to Dark Green Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0267_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart