KEY465192792 Key Organics / BIONET

1-(4-chlorophenyl)-4-[2-(3-fluoropropoxy)benzoyl]-1H-pyrazole

  • MDL number MFCD04124580
  • Cas Number : 956794-46-0
  • Molecular Weight : 358.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9998

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0288_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart