KEY465192797 Key Organics / BIONET

1-(4-chlorophenyl)-3-[2-(5-methoxy-1H-indol-3-yl)ethyl]urea

  • MDL number MFCD01260785
  • Cas Number : 860786-97-6
  • Molecular Weight : 343.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/62888

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart