KEY465192805 Key Organics / BIONET

2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)butanedioic acid

  • MDL number MFCD04124607
  • Cas Number : 26577-34-4
  • Molecular Weight : 249.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/62917

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0849_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart