KEY465192807 Key Organics / BIONET

(2Z)-3-{4-[(Z)-[(4-methoxyphenyl)methylidene]amino]phenyl}prop-2-enoic acid

  • MDL number MFCD04124620
  • Cas Number : 25959-50-6
  • Molecular Weight : 281.311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/62944

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0892_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart