KEY465192810 Key Organics / BIONET

4-(5-methoxy-1,3-benzothiazol-2-yl)aniline

  • MDL number MFCD22375456
  • Cas Number : 162374-77-8
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63007

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart