KEY465192812 Key Organics / BIONET

1-{4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenesulfonyl}piperidin-4-one

  • MDL number MFCD05975053
  • Cas Number : 860787-37-7
  • Molecular Weight : 386.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0811_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart