KEY465192813 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(2,4-difluorophenyl)propanoic acid

  • MDL number MFCD06496115
  • Cas Number : 860787-38-8
  • Molecular Weight : 301.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63032

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart