KEY465192815 Key Organics / BIONET

(2E)-3-{1-[(2-chloro-1,3-thiazol-5-yl)methyl]-1H-indol-3-yl}-2-methanesulfonylprop-2-enenitrile

  • MDL number MFCD05975055
  • Cas Number : 860787-40-2
  • Molecular Weight : 377.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63039

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0819_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart