KEY465192816 Key Organics / BIONET

N'-[(1E)-{1-[(2-chloro-1,3-thiazol-5-yl)methyl]-1H-indol-3-yl}methylidene]pyridine-4-carbohydrazide

  • MDL number MFCD05975056
  • Cas Number : 860787-41-3
  • Molecular Weight : 395.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63040

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0820_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart