KEY465192820 Key Organics / BIONET

methyl 2-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetate

  • MDL number MFCD00457632
  • Cas Number : 134456-94-3
  • Molecular Weight : 198.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0833_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart