KEY465192822 Key Organics / BIONET

1-[5-(phenylamino)-1,2,4-thiadiazol-3-yl]propan-2-one

  • MDL number MFCD00084856
  • Cas Number : 64822-00-0
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63053

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0839_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart