KEY465192823 Key Organics / BIONET

3-{1-[4-methyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-1,2,3-triazol-4-yl}aniline

  • MDL number MFCD06496124
  • Cas Number : 860787-47-9
  • Molecular Weight : 320.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0852_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart