KEY465192829 Key Organics / BIONET

4-chloro-N-[(furan-2-yl)methyl]benzene-1-sulfonamide

  • MDL number MFCD00034421
  • Cas Number : 196813-49-7
  • Molecular Weight : 271.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart