KEY465192832 Key Organics / BIONET

3-(4-chlorobenzenesulfonyl)-10-oxa-3-azatricyclo[5.2.1.0^{1,5}]dec-8-ene

  • MDL number MFCD05975062
  • Cas Number : 65193-52-4
  • Molecular Weight : 311.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63076

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0946_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart