KEY465192833 Key Organics / BIONET

(E)-N'-[3-cyano-4-(4-methylphenyl)-4H-benzo[h]chromen-2-yl]-N,N-dimethylmethanimidamide

  • MDL number MFCD06496125
  • Cas Number : 860787-52-6
  • Molecular Weight : 367.452

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63079

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0952_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart