KEY465192835 Key Organics / BIONET

2-(chloromethyl)-5-(4-iodophenyl)-1,3,4-oxadiazole

  • MDL number MFCD07021344
  • Cas Number : 623907-47-1
  • Molecular Weight : 320.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63104

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12Y-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart