KEY465192839 Key Organics / BIONET

2-chloro-N'-[6-chloro-4-(trifluoromethyl)pyridin-2-yl]benzohydrazide

  • MDL number MFCD27981281
  • Cas Number : 1797764-46-5
  • Molecular Weight : 350.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220 - 222
Purity 97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431123

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12Z-5014_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart