KEY465192843 Key Organics / BIONET

N'-[6-chloro-4-(trifluoromethyl)pyridin-2-yl]-2,6-difluorobenzohydrazide

  • MDL number MFCD27981282
  • Cas Number : 1797988-93-2
  • Molecular Weight : 351.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241 - 243
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12Z-5021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart