KEY465192844 Key Organics / BIONET

N-(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-2-methylbenzene-1-sulfonamide

  • MDL number MFCD11501693
  • Cas Number : 1092344-09-6
  • Molecular Weight : 394.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63138

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12Z-5033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart