KEY465192847 Key Organics / BIONET

(1Z)-2-(4-hydroxy-1,3-thiazol-2-yl)-4,4-dimethyl-1-phenylpent-1-en-3-one

  • MDL number MFCD00138174
  • Cas Number : 320424-51-9
  • Molecular Weight : 287.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63146

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1B-010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart