KEY465192849 Key Organics / BIONET

3-(nitromethyl)-4-phenyl-5-(trifluoromethyl)-4H-1,2,4-triazole

  • MDL number MFCD00138451
  • Cas Number : 320424-54-2
  • Molecular Weight : 272.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63149

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1B-015_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart