KEY465192852 Key Organics / BIONET

dimethyl[(Z)-2-[4-methyl-5-(trifluoromethyl)-4H-1,2,4-triazol-3-yl]-2-nitroethenyl]amine

  • MDL number MFCD00138580
  • Cas Number : 320424-59-7
  • Molecular Weight : 265.196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63157

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1B-026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart