KEY465192857 Key Organics / BIONET

5-nitro-2-[(pyridin-4-yl)methyl]-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD27924205
  • Cas Number : 1797096-04-8
  • Molecular Weight : 281.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 179
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402875

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart