KEY465192861 Key Organics / BIONET

methyl 4-({[(4Z)-3-methyl-5-oxo-1-phenyl-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}amino)benzoate

  • MDL number MFCD00138379
  • Cas Number : 320424-78-0
  • Molecular Weight : 335.363

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63194

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart